Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606027 | ||||||||||||||||||||
Literature Links: |
ADCK5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADCK5 - aarF domain containing kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174922.4 | 3974 | Intron | NP_777582.4 | |||
XM_006716527.2 | 3974 | Intron | XP_006716590.1 | |||
XM_011516907.2 | 3974 | Intron | XP_011515209.1 | |||
XM_011516909.1 | 3974 | Intron | XP_011515211.1 | |||
XM_011516910.1 | 3974 | Intron | XP_011515212.1 | |||
XM_017013174.1 | 3974 | Intron | XP_016868663.1 |
CPSF1 - cleavage and polyadenylation specific factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013291.2 | 3974 | Missense Mutation | CGG,TGG | R,W 1317 | NP_037423.2 | |
XM_006716548.2 | 3974 | Missense Mutation | CGG,TGG | R,W 1314 | XP_006716611.1 | |
XM_006716549.2 | 3974 | Missense Mutation | CGG,TGG | R,W 1239 | XP_006716612.1 | |
XM_006716550.3 | 3974 | Missense Mutation | CGG,TGG | R,W 659 | XP_006716613.1 | |
XM_011516997.1 | 3974 | Missense Mutation | CGG,TGG | R,W 1239 | XP_011515299.1 | |
XM_011516998.1 | 3974 | Missense Mutation | CGG,TGG | R,W 1239 | XP_011515300.1 | |
XM_011516999.1 | 3974 | Missense Mutation | CGG,TGG | R,W 1239 | XP_011515301.1 |
MIR1234 - microRNA 1234 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6849 - microRNA 6849 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR939 - microRNA 939 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |