Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACGCGCGAGAAGCCCTTCTGCTG[C/T]GAGGCCTGCGGGCAGGCGTTCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BREA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BREA2 - breast cancer estrogen-induced apoptosis 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF707 - zinc finger protein 707 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100598.1 | 943 | Silent Mutation | TGC,TGT | C,C 234 | NP_001094068.1 | |
NM_001100599.1 | 943 | Silent Mutation | TGC,TGT | C,C 234 | NP_001094069.1 | |
NM_001288805.1 | 943 | Silent Mutation | TGC,TGT | C,C 234 | NP_001275734.1 | |
NM_001288806.1 | 943 | Silent Mutation | TGC,TGT | C,C 234 | NP_001275735.1 | |
NM_001288807.1 | 943 | Silent Mutation | TGC,TGT | C,C 161 | NP_001275736.1 | |
NM_001288808.1 | 943 | Silent Mutation | TGC,TGT | C,C 198 | NP_001275737.1 | |
NM_001288809.1 | 943 | Silent Mutation | TGC,TGT | C,C 198 | NP_001275738.1 | |
NM_173831.3 | 943 | Silent Mutation | TGC,TGT | C,C 234 | NP_776192.2 | |
XM_005250883.3 | 943 | Silent Mutation | TGC,TGT | C,C 161 | XP_005250940.1 | |
XM_006716542.3 | 943 | Silent Mutation | TGC,TGT | C,C 161 | XP_006716605.1 | |
XM_006716543.3 | 943 | Silent Mutation | TGC,TGT | C,C 161 | XP_006716606.1 | |
XM_011516977.2 | 943 | Silent Mutation | TGC,TGT | C,C 234 | XP_011515279.1 | |
XM_011516979.2 | 943 | Silent Mutation | TGC,TGT | C,C 161 | XP_011515281.1 | |
XM_017013335.1 | 943 | Silent Mutation | TGC,TGT | C,C 161 | XP_016868824.1 |