Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTCCCAGAGGCTGGAGTGGCT[A/G]TCCTAGGCAGGGCTAGGGGAGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS55 - protease, serine 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197020.1 | 346 | Missense Mutation | ATC,GTC | I,V 31 | NP_001183949.1 | |
NM_198464.3 | 346 | Missense Mutation | ATC,GTC | I,V 31 | NP_940866.2 | |
XM_005272372.3 | 346 | Missense Mutation | ATC,GTC | I,V 96 | XP_005272429.2 | |
XM_011543814.2 | 346 | Missense Mutation | ATC,GTC | I,V 96 | XP_011542116.2 | |
XM_017013182.1 | 346 | Missense Mutation | ATC,GTC | I,V 96 | XP_016868671.1 |