Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAGTAGTCGAGGCGGCTGCCG[C/T]AGAAGCGCGGCATCTCCTCCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606396 MIM: 607359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BIN3 - bridging integrator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018688.4 | 742 | Missense Mutation | TAC,TGC | Y,C 187 | NP_061158.1 | |
XM_005273573.3 | 742 | Missense Mutation | TAC,TGC | Y,C 139 | XP_005273630.1 | |
XM_011544586.2 | 742 | Missense Mutation | TAC,TGC | Y,C 133 | XP_011542888.1 | |
XM_011544587.1 | 742 | Intron | XP_011542889.1 |
CCAR2 - cell cycle and apoptosis regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |