Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGCCACAGCCGCCGCCAGGGG[G/T]TGGTGCGCCTGCGTCTGCGGGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP7 - charged multivesicular body protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317899.1 | 779 | Intron | NP_001304828.1 | |||
NM_152272.4 | 779 | Missense Mutation | GTG,TTG | V,L 69 | NP_689485.1 | |
XM_006716415.2 | 779 | Missense Mutation | GTG,TTG | V,L 69 | XP_006716478.1 | |
XM_017013961.1 | 779 | Missense Mutation | GTG,TTG | V,L 69 | XP_016869450.1 | |
XM_017013962.1 | 779 | UTR 5 | XP_016869451.1 | |||
XM_017013963.1 | 779 | Intron | XP_016869452.1 | |||
XM_017013964.1 | 779 | UTR 5 | XP_016869453.1 | |||
XM_017013965.1 | 779 | Intron | XP_016869454.1 |