Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCGCCCGGGAAGTCAACACGGC[G/T]TCGCTGTGCCGCATCGGGCAGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610237 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED30 - mediator complex subunit 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282986.1 | 424 | Silent Mutation | GCG,GCT | A,A 31 | NP_001269915.1 | |
NM_080651.3 | 424 | Silent Mutation | GCG,GCT | A,A 31 | NP_542382.1 | |
XM_011517360.2 | 424 | Silent Mutation | GCG,GCT | A,A 31 | XP_011515662.1 |