Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCTCACCCAGGTCAGAGTTCAA[C/T]AGCTTCTGTGCCATCTCAATCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTK2 - protein tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199649.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1002 | NP_001186578.1 | |
NM_001316342.1 | 3073 | Silent Mutation | CTA,CTG | L,L 892 | NP_001303271.1 | |
NM_005607.4 | 3073 | Silent Mutation | CTA,CTG | L,L 1011 | NP_005598.3 | |
NM_153831.3 | 3073 | Silent Mutation | CTA,CTG | L,L 989 | NP_722560.1 | |
XM_005251003.2 | 3073 | Silent Mutation | CTA,CTG | L,L 1077 | XP_005251060.1 | |
XM_005251012.2 | 3073 | Silent Mutation | CTA,CTG | L,L 1033 | XP_005251069.1 | |
XM_005251013.2 | 3073 | Silent Mutation | CTA,CTG | L,L 920 | XP_005251070.1 | |
XM_006716608.2 | 3073 | Silent Mutation | CTA,CTG | L,L 1033 | XP_006716671.1 | |
XM_006716609.2 | 3073 | Silent Mutation | CTA,CTG | L,L 920 | XP_006716672.1 | |
XM_006716610.2 | 3073 | Silent Mutation | CTA,CTG | L,L 920 | XP_006716673.1 | |
XM_011517194.2 | 3073 | Silent Mutation | CTA,CTG | L,L 1033 | XP_011515496.1 | |
XM_017013651.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1030 | XP_016869140.1 | |
XM_017013652.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1012 | XP_016869141.1 | |
XM_017013653.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1049 | XP_016869142.1 | |
XM_017013654.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1043 | XP_016869143.1 | |
XM_017013655.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1041 | XP_016869144.1 | |
XM_017013656.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1040 | XP_016869145.1 | |
XM_017013657.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1036 | XP_016869146.1 | |
XM_017013658.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1026 | XP_016869147.1 | |
XM_017013659.1 | 3073 | Silent Mutation | CTA,CTG | L,L 968 | XP_016869148.1 | |
XM_017013660.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1005 | XP_016869149.1 | |
XM_017013661.1 | 3073 | Silent Mutation | CTA,CTG | L,L 1005 | XP_016869150.1 | |
XM_017013662.1 | 3073 | Silent Mutation | CTA,CTG | L,L 999 | XP_016869151.1 | |
XM_017013663.1 | 3073 | Silent Mutation | CTA,CTG | L,L 999 | XP_016869152.1 | |
XM_017013664.1 | 3073 | Silent Mutation | CTA,CTG | L,L 998 | XP_016869153.1 | |
XM_017013665.1 | 3073 | Silent Mutation | CTA,CTG | L,L 998 | XP_016869154.1 | |
XM_017013666.1 | 3073 | Silent Mutation | CTA,CTG | L,L 997 | XP_016869155.1 | |
XM_017013667.1 | 3073 | Silent Mutation | CTA,CTG | L,L 997 | XP_016869156.1 | |
XM_017013668.1 | 3073 | Silent Mutation | CTA,CTG | L,L 997 | XP_016869157.1 | |
XM_017013669.1 | 3073 | Silent Mutation | CTA,CTG | L,L 996 | XP_016869158.1 | |
XM_017013670.1 | 3073 | Silent Mutation | CTA,CTG | L,L 995 | XP_016869159.1 | |
XM_017013671.1 | 3073 | Silent Mutation | CTA,CTG | L,L 992 | XP_016869160.1 | |
XM_017013672.1 | 3073 | Silent Mutation | CTA,CTG | L,L 992 | XP_016869161.1 | |
XM_017013673.1 | 3073 | Silent Mutation | CTA,CTG | L,L 989 | XP_016869162.1 | |
XM_017013674.1 | 3073 | Silent Mutation | CTA,CTG | L,L 984 | XP_016869163.1 | |
XM_017013675.1 | 3073 | Silent Mutation | CTA,CTG | L,L 984 | XP_016869164.1 | |
XM_017013676.1 | 3073 | Silent Mutation | CTA,CTG | L,L 971 | XP_016869165.1 | |
XM_017013677.1 | 3073 | Silent Mutation | CTA,CTG | L,L 969 | XP_016869166.1 | |
XM_017013678.1 | 3073 | Silent Mutation | CTA,CTG | L,L 953 | XP_016869167.1 | |
XM_017013679.1 | 3073 | Silent Mutation | CTA,CTG | L,L 948 | XP_016869168.1 | |
XM_017013680.1 | 3073 | Silent Mutation | CTA,CTG | L,L 932 | XP_016869169.1 | |
XM_017013681.1 | 3073 | Silent Mutation | CTA,CTG | L,L 917 | XP_016869170.1 | |
XM_017013682.1 | 3073 | Silent Mutation | CTA,CTG | L,L 913 | XP_016869171.1 | |
XM_017013683.1 | 3073 | Silent Mutation | CTA,CTG | L,L 892 | XP_016869172.1 | |
XM_017013684.1 | 3073 | Silent Mutation | CTA,CTG | L,L 886 | XP_016869173.1 | |
XM_017013685.1 | 3073 | Silent Mutation | CTA,CTG | L,L 885 | XP_016869174.1 | |
XM_017013686.1 | 3073 | Silent Mutation | CTA,CTG | L,L 879 | XP_016869175.1 | |
XM_017013687.1 | 3073 | Silent Mutation | CTA,CTG | L,L 871 | XP_016869176.1 | |
XM_017013688.1 | 3073 | Silent Mutation | CTA,CTG | L,L 856 | XP_016869177.1 | |
XM_017013689.1 | 3073 | Silent Mutation | CTA,CTG | L,L 856 | XP_016869178.1 | |
XM_017013690.1 | 3073 | Silent Mutation | CTA,CTG | L,L 850 | XP_016869179.1 |