Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGCAGAGGACGCGGCGGATGCG[A/G]CAGAGGCAGCGTCTTCAAGTGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123980 MIM: 610210 MIM: 611885 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYC1 - cytochrome c1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAF1 - MAF1 homolog, negative regulator of RNA polymerase III | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHARPIN - SHANK associated RH domain interactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030974.3 | 1176 | Missense Mutation | GCC,GTC | A,V 231 | NP_112236.3 | |
XM_017013887.1 | 1176 | Missense Mutation | GCC,GTC | A,V 231 | XP_016869376.1 | |
XM_017013888.1 | 1176 | Missense Mutation | GCC,GTC | A,V 231 | XP_016869377.1 |
WDR97 - WD repeat domain 97 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |