Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTGTTCCCTGCAGCTGCAGTCC[A/G]TTTGGATAGACTCAGACTTAGGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
XKR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XKR5 - XK related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289973.1 | 2579 | Missense Mutation | NP_001276902.1 | |||
NM_207411.4 | 2579 | Missense Mutation | NP_997294.3 | |||
XM_011534750.2 | 2579 | Missense Mutation | XP_011533052.1 | |||
XM_017013401.1 | 2579 | Missense Mutation | XP_016868890.1 | |||
XM_017013402.1 | 2579 | Missense Mutation | XP_016868891.1 |