Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCAGTAACAATTCACCGTCGTC[C/T]TCTTCTTCCCCGACTTCTTCCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNKS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNKS - tankyrase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003747.2 | 1104 | Silent Mutation | TCC,TCT | S,S 131 | NP_003738.2 | |
XM_006716263.3 | 1104 | Silent Mutation | TCC,TCT | S,S 131 | XP_006716326.1 | |
XM_011543845.2 | 1104 | Silent Mutation | TCC,TCT | S,S 131 | XP_011542147.1 | |
XM_011543846.2 | 1104 | Silent Mutation | TCC,TCT | S,S 131 | XP_011542148.1 | |
XM_011543847.2 | 1104 | Silent Mutation | TCC,TCT | S,S 131 | XP_011542149.1 |