Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGTTTTGTCCCGCAGTATAGAA[C/T]ATTCCATTTAGATTGGAGGGGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601667 | ||||||||||||||||||||
Literature Links: |
ANGPT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGPT1 - angiopoietin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146.4 | 1059 | Missense Mutation | ATA,ATG | I,M 460 | NP_001137.2 | |
NM_001199859.2 | 1059 | Missense Mutation | ATA,ATG | I,M 459 | NP_001186788.1 | |
NM_001314051.1 | 1059 | Missense Mutation | ATA,ATG | I,M 260 | NP_001300980.1 |