Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCTCTATCCATCTCTCGAAGA[C/G]TTGAAGGTAGACAAAGTAATTCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602217 | ||||||||||||||||||||
Literature Links: |
SDCBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDCBP - syndecan binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007067.1 | 1549 | Intron | NP_001007068.1 | |||
NM_001007068.1 | 1549 | Intron | NP_001007069.1 | |||
NM_001007069.1 | 1549 | Intron | NP_001007070.1 | |||
NM_001007070.1 | 1549 | Intron | NP_001007071.1 | |||
NM_005625.3 | 1549 | Intron | NP_005616.2 | |||
XM_011517571.2 | 1549 | Missense Mutation | GAC,GAG | D,E 29 | XP_011515873.2 | |
XM_017013727.1 | 1549 | Missense Mutation | GAC,GAG | D,E 30 | XP_016869216.1 | |
XM_017013728.1 | 1549 | Missense Mutation | GAC,GAG | D,E 30 | XP_016869217.1 |