Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATTCCCTACCACAAACTCGCC[A/G]ACCTGCGCTACCTGAGCCGCGGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603455 | ||||||||||||||||||||
Literature Links: |
LOC101929709 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929709 - uncharacterized LOC101929709 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIPK2 - receptor interacting serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003821.5 | 336 | Missense Mutation | AAC,GAC | N,D 20 | NP_003812.1 | |
XM_005251092.3 | 336 | UTR 5 | XP_005251149.1 | |||
XM_011517357.2 | 336 | Intron | XP_011515659.1 |