Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCATGATGCTGTACATAAGGCCC[A/G]TTTCTTTCATTGGTAACATCAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600899 | ||||||||||||||||||||
Literature Links: |
PRKDC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRKDC - protein kinase, DNA-activated, catalytic polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081640.1 | 11894 | Missense Mutation | ACG,ATG | T,M 3946 | NP_001075109.1 | |
NM_006904.6 | 11894 | Missense Mutation | ACG,ATG | T,M 3977 | NP_008835.5 |