Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCCCGTCCGCTCTCATAGTCCG[C/T]GATCACCTGTGGCTTCTCATTGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605107 | ||||||||||||||||||||
Literature Links: |
EDF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EDF1 - endothelial differentiation related factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281297.1 | 343 | Missense Mutation | ACG,GCG | T,A 107 | NP_001268226.1 | |
NM_001281298.1 | 343 | Missense Mutation | ACG,GCG | T,A 86 | NP_001268227.1 | |
NM_001281299.1 | 343 | Missense Mutation | ACG,GCG | T,A 89 | NP_001268228.1 | |
NM_003792.3 | 343 | Missense Mutation | ACG,GCG | T,A 107 | NP_003783.1 | |
NM_153200.2 | 343 | Missense Mutation | ACG,GCG | T,A 107 | NP_694880.1 |
MAMDC4 - MAM domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |