Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCATCCAGGGGCTGAGAGACGG[C/T]GGGACGCTGGGGCAGGGCACACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606249 | ||||||||||||||||||||
Literature Links: |
C9orf129 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf129 - chromosome 9 open reading frame 129 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098808.1 | 933 | Missense Mutation | CAC,CGC | H,R 190 | NP_001092278.1 | |
XM_006717117.3 | 933 | Missense Mutation | CAC,CGC | H,R 190 | XP_006717180.1 |