Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGGCAGCTGATGGCATAAAAAT[A/G]TCTTCATACTTGTAAATACTGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRRS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRRS1L - ferric chelate reductase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011518453.2 | 1320 | Silent Mutation | GAC,GAT | D,D 196 | XP_011516755.1 | |
XM_011518454.2 | 1320 | Silent Mutation | GAC,GAT | D,D 196 | XP_011516756.1 |