Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGTCTCCTCACAGCTGGGCCC[C/T]GGGTAAGTTGGAAATTAACTTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF79 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF79 - zinc finger protein 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286696.1 | 624 | Missense Mutation | CGG,TGG | R,W 11 | NP_001273625.1 | |
NM_001286697.1 | 624 | Missense Mutation | CGG,TGG | R,W 11 | NP_001273626.1 | |
NM_001286698.1 | 624 | UTR 5 | NP_001273627.1 | |||
NM_001322260.1 | 624 | Missense Mutation | CGG,TGG | R,W 11 | NP_001309189.1 | |
NM_007135.2 | 624 | Missense Mutation | CGG,TGG | R,W 35 | NP_009066.2 | |
XM_006717279.3 | 624 | Missense Mutation | CGG,TGG | R,W 11 | XP_006717342.1 |