Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCAGATGGCCTCCAGGCCAAA[C/G]ATGGGTACTTTGCTGGAAGAAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609312 MIM: 604455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBH - dopamine beta-hydroxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DBH-AS1 - DBH antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SARDH - sarcosine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134707.1 | 2804 | Missense Mutation | ATC,ATG | I,M 836 | NP_001128179.1 | |
NM_007101.3 | 2804 | Missense Mutation | ATC,ATG | I,M 836 | NP_009032.2 | |
XM_017014367.1 | 2804 | Missense Mutation | ATC,ATG | I,M 836 | XP_016869856.1 | |
XM_017014368.1 | 2804 | Intron | XP_016869857.1 |