Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCACCTAAACTCTGTTTCTTCTG[A/C]TTTGGTGGCTCCTTTTGGTGCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615231 | ||||||||||||||||||||
Literature Links: |
RC3H2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RC3H2 - ring finger and CCCH-type domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100588.1 | 3583 | Silent Mutation | NP_001094058.1 | |||
NM_018835.2 | 3583 | Intron | NP_061323.2 |