Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGAGACGGACAAACTTTTTGTC[T/C]TCAGACGAGGGAGCGTTTTGTAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604366 | ||||||||||||||||||||
Literature Links: |
DNAI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAI1 - dynein axonemal intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281428.1 | 214 | UTR 5 | NP_001268357.1 | |||
NM_012144.3 | 214 | UTR 5 | NP_036276.1 | |||
XM_006716758.3 | 214 | Intron | XP_006716821.1 | |||
XM_011517846.1 | 214 | UTR 5 | XP_011516148.1 | |||
XM_011517847.2 | 214 | UTR 5 | XP_011516149.1 | |||
XM_011517848.1 | 214 | UTR 5 | XP_011516150.1 | |||
XM_011517849.1 | 214 | UTR 5 | XP_011516151.1 | |||
XM_011517850.2 | 214 | UTR 5 | XP_011516152.1 | |||
XM_017014625.1 | 214 | UTR 5 | XP_016870114.1 |
FAM219A - family with sequence similarity 219 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184940.1 | 214 | Intron | NP_001171869.1 | |||
NM_001184941.1 | 214 | Intron | NP_001171870.1 | |||
NM_001184942.1 | 214 | Intron | NP_001171871.1 | |||
NM_001184943.1 | 214 | Intron | NP_001171872.1 | |||
NM_001184945.1 | 214 | Intron | NP_001171874.1 | |||
NM_147202.1 | 214 | Intron | NP_671735.1 | |||
XM_005251402.4 | 214 | Intron | XP_005251459.1 | |||
XM_011517797.2 | 214 | Intron | XP_011516099.1 | |||
XM_011517798.2 | 214 | Intron | XP_011516100.1 | |||
XM_011517799.2 | 214 | Intron | XP_011516101.1 |