Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTCCTCTTTAAAGTTACTGCA[A/G]TGCTCAATCACCTTGCTGGAGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100506100 - uncharacterized LOC100506100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZDHHC12 - zinc finger DHHC-type containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZER1 - zyg-11 related cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006336.3 | 2499 | Silent Mutation | CAC,CAT | H,H 753 | NP_006327.2 | |
XM_005251645.2 | 2499 | Silent Mutation | CAC,CAT | H,H 753 | XP_005251702.1 | |
XM_005251646.2 | 2499 | Silent Mutation | CAC,CAT | H,H 751 | XP_005251703.1 | |
XM_011518122.2 | 2499 | Silent Mutation | CAC,CAT | H,H 753 | XP_011516424.1 | |
XM_017014185.1 | 2499 | Silent Mutation | CAC,CAT | H,H 753 | XP_016869674.1 | |
XM_017014186.1 | 2499 | Silent Mutation | CAC,CAT | H,H 751 | XP_016869675.1 | |
XM_017014187.1 | 2499 | Silent Mutation | CAC,CAT | H,H 751 | XP_016869676.1 | |
XM_017014188.1 | 2499 | Silent Mutation | CAC,CAT | H,H 751 | XP_016869677.1 | |
XM_017014189.1 | 2499 | Silent Mutation | CAC,CAT | H,H 690 | XP_016869678.1 | |
XM_017014190.1 | 2499 | Silent Mutation | CAC,CAT | H,H 690 | XP_016869679.1 |