Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGATCTGCAGAGCCATGGAGGC[A/G]CAGGGGAGCTGGGCCAGGGTGTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605172 | ||||||||||||||||||||
Literature Links: |
PTGES PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGES - prostaglandin E synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004878.4 | 825 | Silent Mutation | TGC,TGT | C,C 137 | NP_004869.1 | |
XM_011519210.2 | 825 | Silent Mutation | TGC,TGT | C,C 165 | XP_011517512.1 |