Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTAGCCCTGTGTCAGGTGAGG[C/G]TGGGACCCAGCTGTCCTCAGTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607387 | ||||||||||||||||||||
Literature Links: |
DCTN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCTN3 - dynactin subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPP25L - ribonuclease P/MRP subunit p25 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148178.2 | 585 | Missense Mutation | CCC,GCC | P,A 99 | NP_680544.1 | |
NM_148179.2 | 585 | Missense Mutation | CCC,GCC | P,A 99 | NP_680545.1 |