Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCAGGAGCTCCGGCCCGGACC[C/T]CACGCGATGGAGGAAGCAGCAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600777 | ||||||||||||||||||||
Literature Links: |
CFAP77 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFAP77 - cilia and flagella associated protein 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282957.1 | 261 | Missense Mutation | CCC,CTC | P,L 11 | NP_001269886.1 | |
NM_207417.2 | 261 | Missense Mutation | CCC,CTC | P,L 11 | NP_997300.1 | |
XM_011518670.2 | 261 | Missense Mutation | CCC,CTC | P,L 11 | XP_011516972.1 | |
XM_011518671.1 | 261 | Missense Mutation | CCC,CTC | P,L 11 | XP_011516973.1 | |
XM_017014711.1 | 261 | Missense Mutation | CCC,CTC | P,L 11 | XP_016870200.1 |
TTF1 - transcription termination factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |