Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACGCTTTTCCAGCTCCTTCTTCT[G/T]TTCCTGAGCTAGCTCCTCTTTCGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD3 - bromodomain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007371.3 | 2529 | Missense Mutation | AAG,CAG | K,Q 664 | NP_031397.1 | |
XM_006717291.2 | 2529 | Missense Mutation | AAG,CAG | K,Q 664 | XP_006717354.1 | |
XM_011519052.2 | 2529 | Missense Mutation | AAG,CAG | K,Q 664 | XP_011517354.1 | |
XM_017015165.1 | 2529 | Missense Mutation | AAG,CAG | K,Q 663 | XP_016870654.1 | |
XM_017015166.1 | 2529 | Missense Mutation | AAG,CAG | K,Q 343 | XP_016870655.1 |
LINC00094 - long intergenic non-protein coding RNA 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |