Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGCTGACATTTGAGGATGTGGC[C/T]GTGTTTTTTACCCAGGAGGAGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611854 MIM: 603132 | ||||||||||||||||||||
Literature Links: |
MRPL50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL50 - mitochondrial ribosomal protein L50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF189 - zinc finger protein 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278231.1 | 514 | Intron | NP_001265160.1 | |||
NM_001278232.1 | 514 | Intron | NP_001265161.1 | |||
NM_001278240.1 | 514 | Intron | NP_001265169.1 | |||
NM_003452.3 | 514 | Silent Mutation | GCC,GCT | A,A 20 | NP_003443.2 | |
NM_197977.2 | 514 | Intron | NP_932094.1 | |||
XM_006717280.3 | 514 | UTR 5 | XP_006717343.1 | |||
XM_006717281.3 | 514 | Intron | XP_006717344.1 | |||
XM_011518998.2 | 514 | Intron | XP_011517300.1 | |||
XM_011518999.2 | 514 | Intron | XP_011517301.1 | |||
XM_017015121.1 | 514 | Intron | XP_016870610.1 |