Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGGCTCTCCTATGTGGGAAACA[A/G]TTTTGGGACTGGAGCCCTCAAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
POLR1E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR1E - RNA polymerase I subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282766.1 | 331 | UTR 5 | NP_001269695.1 | |||
NM_022490.2 | 331 | Missense Mutation | AAT,AGT | N,S 73 | NP_071935.1 | |
XM_005251547.2 | 331 | Missense Mutation | AAT,AGT | N,S 135 | XP_005251604.1 |