Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGTGCTTCATGCCCGAGACCCC[A/G]CGCTTCCTGCTGACTCAGCACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605245 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GARNL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GARNL3 - GTPase activating Rap/RanGAP domain like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC2A8 - solute carrier family 2 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271711.1 | 711 | Silent Mutation | CCA,CCG | P,P 206 | NP_001258640.1 | |
NM_001271712.1 | 711 | Silent Mutation | CCA,CCG | P,P 43 | NP_001258641.1 | |
NM_014580.4 | 711 | Silent Mutation | CCA,CCG | P,P 206 | NP_055395.2 | |
XM_006717084.3 | 711 | Silent Mutation | CCA,CCG | P,P 206 | XP_006717147.1 | |
XM_011518602.2 | 711 | Silent Mutation | CCA,CCG | P,P 206 | XP_011516904.1 | |
XM_011518603.2 | 711 | Silent Mutation | CCA,CCG | P,P 71 | XP_011516905.1 | |
XM_011518604.2 | 711 | Silent Mutation | CCA,CCG | P,P 71 | XP_011516906.1 | |
XM_017014653.1 | 711 | Silent Mutation | CCA,CCG | P,P 43 | XP_016870142.1 |