Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGGCACCTGGTCCTTCTCTGT[A/G]TCCTTTCTTCCCTTGGCCTCCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611053 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM166B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM166B - family with sequence similarity 166 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099951.3 | 494 | Silent Mutation | GAC,GAT | D,D 141 | NP_001093421.1 | |
NM_001164310.2 | 494 | Silent Mutation | GAC,GAT | D,D 141 | NP_001157782.1 | |
NM_001287238.1 | 494 | Silent Mutation | GAC,GAT | D,D 141 | NP_001274167.1 | |
NM_001287239.1 | 494 | Silent Mutation | GAC,GAT | D,D 141 | NP_001274168.1 | |
XM_011518028.2 | 494 | Silent Mutation | GAC,GAT | D,D 136 | XP_011516330.1 |
RUSC2 - RUN and SH3 domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |