Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGCGGCTGCCCGTACTTGGGC[C/T]GCCACATGAGGACCTGGGCATCGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM78A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM78A - family with sequence similarity 78 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033387.3 | 1143 | Missense Mutation | CAG,CGG | Q,R 268 | NP_203745.2 | |
XM_011518567.1 | 1143 | Missense Mutation | CAG,CGG | Q,R 219 | XP_011516869.1 | |
XM_011518568.2 | 1143 | Missense Mutation | CAG,CGG | Q,R 185 | XP_011516870.1 |