Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGGAAGACTTTGTCCAGAGAGA[A/G]TTTGCCGAGCTCTGATCTCATGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614730 MIM: 608292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM214B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM214B - family with sequence similarity 214 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGO - phosphatidylinositol glycan anchor biosynthesis class O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STOML2 - stomatin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287031.1 | 411 | Missense Mutation | CTC,TTC | L,F 141 | NP_001273960.1 | |
NM_001287032.1 | 411 | Missense Mutation | CTC,TTC | L,F 90 | NP_001273961.1 | |
NM_001287033.1 | 411 | Missense Mutation | CTC,TTC | L,F 95 | NP_001273962.1 | |
NM_013442.2 | 411 | Missense Mutation | CTC,TTC | L,F 141 | NP_038470.1 |