Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTGCCAGCCACCAGTGCGTTG[C/T]GCCAGAATTTCTCCGTAATGTAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600047 MIM: 602030 MIM: 605798 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA2 - ATP binding cassette subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C9orf139 - chromosome 9 open reading frame 139 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207511.2 | 1577 | Intron | NP_997394.1 | |||
XM_017014717.1 | 1577 | Intron | XP_016870206.1 |
FUT7 - fucosyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004479.3 | 1577 | Missense Mutation | NP_004470.1 |
NPDC1 - neural proliferation, differentiation and control 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |