Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGCAGTTGGGCTCCGCGCCGTG[C/G]AGCAGCAGCAGCTCCGCCACGCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600431 MIM: 613149 | ||||||||||||||||||||
Literature Links: |
CDKN2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKN2B - cyclin dependent kinase inhibitor 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004936.3 | 684 | Silent Mutation | CTC,CTG | L,L 67 | NP_004927.2 | |
NM_078487.2 | 684 | UTR 3 | NP_511042.1 |
CDKN2B-AS1 - CDKN2B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |