Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATAGGCTTATTAAGTTACACAT[C/T]TAAAAAGTCATTAGAACATCTCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf72 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf72 - chromosome 9 open reading frame 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256054.2 | 1553 | Silent Mutation | TAA,TAG | *,* 482 | NP_001242983.1 | |
NM_018325.4 | 1553 | Silent Mutation | TAA,TAG | *,* 482 | NP_060795.1 | |
NM_145005.6 | 1553 | Intron | NP_659442.2 |