Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTCCGTCTGCAGACGGCAAC[C/G]GGGGCCTCTACTGCGCGCTGGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604346 MIM: 612057 | ||||||||||||||||||||
Literature Links: |
MAN1B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAN1B1 - mannosidase alpha class 1B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN1B1-AS1 - MAN1B1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAPCD2 - suppressor APC domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UAP1L1 - UDP-N-acetylglucosamine pyrophosphorylase 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207309.2 | 737 | Missense Mutation | CGG,GGG | R,G 227 | NP_997192.2 | |
XM_006717317.3 | 737 | Missense Mutation | CGG,GGG | R,G 227 | XP_006717380.1 | |
XM_011519182.2 | 737 | Missense Mutation | CGG,GGG | R,G 60 | XP_011517484.1 |