Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGAGGCCAGCACTGTGGGCTGG[C/T]GACAGAGTGGGCACAGCAGGCGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607795 | ||||||||||||||||||||
Literature Links: |
PRPF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRPF4 - pre-mRNA processing factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF183 - ring finger protein 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145051.3 | 1890 | Missense Mutation | CAC,CGC | H,R 60 | NP_659488.2 | |
XM_006716952.2 | 1890 | Missense Mutation | CAC,CGC | H,R 60 | XP_006717015.1 | |
XM_011518218.1 | 1890 | Missense Mutation | CAC,CGC | H,R 60 | XP_011516520.1 | |
XM_011518219.1 | 1890 | Missense Mutation | CAC,CGC | H,R 60 | XP_011516521.1 | |
XM_011518220.1 | 1890 | Missense Mutation | CAC,CGC | H,R 60 | XP_011516522.1 | |
XM_017014268.1 | 1890 | Missense Mutation | CAC,CGC | H,R 60 | XP_016869757.1 |