Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCATGAACCCCGACTACACCAAG[C/G]TGACCATGGACCTTGGCAGCACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928438 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928438 - uncharacterized LOC101928438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR4A3 - nuclear receptor subfamily 4 group A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006981.3 | 773 | Missense Mutation | CTG,GTG | L,V 38 | NP_008912.2 | |
NM_173199.2 | 773 | Missense Mutation | CTG,GTG | L,V 38 | NP_775291.1 | |
NM_173200.2 | 773 | Missense Mutation | CTG,GTG | L,V 49 | NP_775292.1 | |
XM_017015162.1 | 773 | Missense Mutation | CTG,GTG | L,V 38 | XP_016870651.1 |