Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGCGGCGGCGGCGGCGGCTGC[A/G]GTAGCAGCAGCAGCTACTGCAGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616130 | ||||||||||||||||||||
Literature Links: |
LURAP1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LURAP1L - leucine rich adaptor protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203403.1 | 872 | Missense Mutation | AGT,GGT | S,G 57 | NP_981948.1 | |
XM_005251443.2 | 872 | Missense Mutation | AGT,GGT | S,G 57 | XP_005251500.1 |
LURAP1L-AS1 - LURAP1L antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |