Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGATTTTCTCTTCCAAAAGAT[C/G]CTTCTTGTTCAAGAATAAAATCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604397 MIM: 605978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNA14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNA14 - G protein subunit alpha 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004297.3 | 1340 | Missense Mutation | CAT,GAT | H,D 273 | NP_004288.1 | |
XM_011519224.1 | 1340 | Missense Mutation | CAT,GAT | H,D 155 | XP_011517526.1 | |
XM_017015341.1 | 1340 | Missense Mutation | CAT,GAT | H,D 219 | XP_016870830.1 |
VPS13A - vacuolar protein sorting 13 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |