Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAATTGTCCAATTGGAGATGGAG[A/G]TTTAGAAAGTGAGTTTAAATCTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CEP78 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP78 - centrosomal protein 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098802.1 | 780 | Missense Mutation | GAT,GGT | D,G 164 | NP_001092272.1 | |
NM_032171.1 | 780 | Missense Mutation | GAT,GGT | D,G 164 | NP_115547.1 | |
XM_005252263.4 | 780 | Missense Mutation | GAT,GGT | D,G 164 | XP_005252320.1 | |
XM_005252265.2 | 780 | Missense Mutation | GAT,GGT | D,G 164 | XP_005252322.1 | |
XM_005252266.4 | 780 | Missense Mutation | GAT,GGT | D,G 77 | XP_005252323.1 | |
XM_017015192.1 | 780 | Missense Mutation | GAT,GGT | D,G 164 | XP_016870681.1 | |
XM_017015193.1 | 780 | Missense Mutation | GAT,GGT | D,G 164 | XP_016870682.1 | |
XM_017015194.1 | 780 | Missense Mutation | GAT,GGT | D,G 164 | XP_016870683.1 | |
XM_017015195.1 | 780 | Missense Mutation | GAT,GGT | D,G 164 | XP_016870684.1 | |
XM_017015196.1 | 780 | Missense Mutation | GAT,GGT | D,G 77 | XP_016870685.1 |