Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGGGCCTCGTCCACCTGCAGAG[C/T]GTCACAGTCCCCCAGCAGCTCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602337 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ROR2 - receptor tyrosine kinase like orphan receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318204.1 | 1696 | Intron | NP_001305133.1 | |||
NM_004560.3 | 1696 | Missense Mutation | ACT,GCT | T,A 931 | NP_004551.2 | |
XM_005252008.4 | 1696 | Missense Mutation | ACT,GCT | T,A 791 | XP_005252065.1 | |
XM_005252009.3 | 1696 | Missense Mutation | ACT,GCT | T,A 530 | XP_005252066.1 | |
XM_006717121.3 | 1696 | Missense Mutation | ACT,GCT | T,A 791 | XP_006717184.1 | |
XM_017014762.1 | 1696 | Missense Mutation | ACT,GCT | T,A 928 | XP_016870251.1 | |
XM_017014763.1 | 1696 | Missense Mutation | ACT,GCT | T,A 791 | XP_016870252.1 |