Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGACACCATCTTCTCCTCACAC[A/G]TGATTTTGGTGTCCTTGAGGATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASTN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASTN2 - astrotactin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184734.1 | 1351 | Missense Mutation | ACG,ATG | T,M 372 | NP_001171663.1 | |
NM_001184735.1 | 1351 | Intron | NP_001171664.1 | |||
NM_014010.4 | 1351 | Missense Mutation | ACG,ATG | T,M 1269 | NP_054729.3 | |
NM_198186.3 | 1351 | Missense Mutation | ACG,ATG | T,M 421 | NP_937829.3 | |
NM_198187.3 | 1351 | Missense Mutation | ACG,ATG | T,M 372 | NP_937830.3 | |
NM_198188.2 | 1351 | Missense Mutation | ACG,ATG | T,M 372 | NP_937831.1 |