Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCACCTCCAGGTTGGGGATGCCT[C/T]AGACTTGTGATGGGACTGGGCAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300007 | ||||||||||||||||||||
Literature Links: |
IL9R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL9R - interleukin 9 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002186.2 | Intron | NP_002177.2 | ||||
NM_176786.1 | Intron | NP_789743.2 | ||||
XM_011531151.2 | Intron | XP_011529453.1 | ||||
XM_011531152.2 | Intron | XP_011529454.1 | ||||
XM_011531154.2 | Intron | XP_011529456.1 | ||||
XM_011531155.2 | Intron | XP_011529457.1 | ||||
XM_011531156.2 | Intron | XP_011529458.1 | ||||
XM_011531157.2 | Intron | XP_011529459.1 | ||||
XM_017029495.1 | Intron | XP_016884984.1 | ||||
XM_017029496.1 | Intron | XP_016884985.1 | ||||
XM_017029497.1 | Intron | XP_016884986.1 | ||||
XM_017029498.1 | Intron | XP_016884987.1 | ||||
XM_017029499.1 | Intron | XP_016884988.1 | ||||
XM_017029500.1 | Intron | XP_016884989.1 | ||||
XM_017029501.1 | Intron | XP_016884990.1 | ||||
XM_017029502.1 | Intron | XP_016884991.1 | ||||
XM_017029503.1 | Intron | XP_016884992.1 | ||||
XM_017029504.1 | Intron | XP_016884993.1 | ||||
XM_017029505.1 | Intron | XP_016884994.1 | ||||
XM_017029506.1 | Intron | XP_016884995.1 |