Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCCTTCAGGGTGCGCACCGGC[C/G]TCGGAGCGCACGGACGTGGTCCTGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 300525 | |||||||||||||||||||||||
Literature Links: |
P2RY8 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
P2RY8 - purinergic receptor P2Y8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178129.4 | 1373 | Missense Mutation | GAC,GAG | D,E 336 | NP_835230.1 | |
XM_005274429.3 | 1373 | Missense Mutation | GAC,GAG | D,E 336 | XP_005274486.1 | |
XM_006724443.3 | 1373 | Missense Mutation | GAC,GAG | D,E 525 | XP_006724506.2 | |
XM_011546178.2 | 1373 | Missense Mutation | GAC,GAG | D,E 336 | XP_011544480.1 | |
XM_011546179.2 | 1373 | Missense Mutation | GAC,GAG | D,E 336 | XP_011544481.1 |