Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACCTCTGACCAGTCGCTTGGGT[A/G]TGTGTCTGGCCCATATACATCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300357 MIM: 400023 | ||||||||||||||||||||
Literature Links: |
CRLF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRLF2 - cytokine receptor-like factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012288.2 | 684 | Missense Mutation | CAC,TAC | H,Y 87 | NP_001012288.2 | |
NM_022148.3 | 684 | Missense Mutation | CAC,TAC | H,Y 199 | NP_071431.2 | |
XM_011546181.2 | 684 | Missense Mutation | CAC,TAC | H,Y 198 | XP_011544483.1 | |
XM_011546182.2 | 684 | Missense Mutation | ACA,ATA | T,I 174 | XP_011544484.1 |