Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
ACCAGTCTAGCGGTCCTCCCGGACC[A/G]GCCGGATCTTCATCTCAGAGAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613357 MIM: 609795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIBCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FIBCD1 - fibrinogen C domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145106.1 | 1474 | Missense Mutation | NP_001138578.1 | |||
NM_032843.4 | 1474 | Missense Mutation | NP_116232.3 |
QRFP - pyroglutamylated RFamide peptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |