Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCGTACAGCTGCAGGGGGAAACC[A/G]AGTCCGCCGGGCCCAGCCTTGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRCT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRCT1 - histidine rich carboxyl terminus 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039792.1 | 251 | Missense Mutation | CAA,CGA | Q,R 52 | NP_001034881.1 |
LINC00961 - long intergenic non-protein coding RNA 961 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |