Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTCTCTCTCTCCAGTTCCTGC[C/T]GGAGTGTCTCTGCACTAAAAAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610340 MIM: 604574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPB41L4B - erythrocyte membrane protein band 4.1 like 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018424.3 | 2786 | Intron | NP_060894.2 | |||
NM_019114.4 | 2786 | Missense Mutation | CAG,CGG | Q,R 883 | NP_061987.3 | |
XM_011518790.1 | 2786 | Missense Mutation | CAG,CGG | Q,R 924 | XP_011517092.1 | |
XM_011518791.2 | 2786 | Missense Mutation | CAG,CGG | Q,R 921 | XP_011517093.1 | |
XM_011518792.2 | 2786 | Missense Mutation | CAG,CGG | Q,R 847 | XP_011517094.1 | |
XM_011518794.2 | 2786 | Missense Mutation | CAG,CGG | Q,R 824 | XP_011517096.1 | |
XM_017014813.1 | 2786 | Missense Mutation | CAG,CGG | Q,R 886 | XP_016870302.1 | |
XM_017014814.1 | 2786 | Missense Mutation | CAG,CGG | Q,R 844 | XP_016870303.1 | |
XM_017014815.1 | 2786 | Missense Mutation | CAG,CGG | Q,R 821 | XP_016870304.1 |
FRRS1L - ferric chelate reductase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |