Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606946 MIM: 138249 MIM: 613505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANAPC2 - anaphase promoting complex subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRIN1 - glutamate ionotropic receptor NMDA type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000832.6 | 604 | Intron | NP_000823.4 | |||
NM_001185090.1 | 604 | Intron | NP_001172019.1 | |||
NM_001185091.1 | 604 | Intron | NP_001172020.1 | |||
NM_007327.3 | 604 | Intron | NP_015566.1 | |||
NM_021569.3 | 604 | Intron | NP_067544.1 | |||
XM_005266071.3 | 604 | Intron | XP_005266128.1 | |||
XM_005266072.3 | 604 | Intron | XP_005266129.1 | |||
XM_005266073.4 | 604 | Intron | XP_005266130.1 | |||
XM_011518583.2 | 604 | Intron | XP_011516885.1 |
LRRC26 - leucine rich repeat containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013653.2 | 604 | Missense Mutation | CGC,TGC | R,C 170 | NP_001013675.1 |
MIR3621 - microRNA 3621 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM210 - transmembrane protein 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |